missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH1 Mouse anti-Human, Clone: ZM001, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen 393200
This item is not returnable.
View return policy
Description
MLH1 is a DNA mismatch repair protein. The repair of mismatch DNA is essential to maintaining the integrity of genetic information over time. An alteration of microsatellite repeats is the result of slippage owing to strand misalignment during DNA replication and is referred to as microsatellite instability (MSI). These defects in DNA repair pathways have been related to human carcinogenesis. The importance of mismatch repair genes became apparent with the identification of the genetic basis for hereditary nonpolyposis colon cancer (HNPCC). MSHS2 is involved in the initial cognition of mismatch nucleotides during the replication mismatch repair process. It is thought that after MSH2 binds to a mismatched DNA duplex it is joined by a heterodimer of MLH1 and PMSH, which together help facilitate the later steps in mismatch repair.
Specifications
| MLH1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| MLH1 | |
| COCA2, FCC2, HNPCC, HNPCC2, hMLH1, DNA mismatch repair protein Mlh1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 4292 | |
| -20°C | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| ZM001 | |
| Unconjugated | |
| P40692 | |
| MLH1 | |
| Recombit protein derived from the C-terminal region of the human MLH1 protein | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction