Learn More
MIP-1b Mouse anti-Human, V450, Clone: D21-1351, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561282
Description
The D21-1351 monoclonal antibody specifically binds to the human CC chemokine, MIP-1β (macrophage inflammatory protein-1β). Human MIP-1β shares approximately 75% homology with mouse MIP-1β at the amino acid level. Expression of MIP-1β in human peripheral blood cells is induced by proinflammatory and mitogenic stimuli. MIP-1βis a chemoattractant for monocytes and lymphocytes. Human MIP-1β
The antibody is conjugated to BD Horizon™ V450, which has been developed for use in multicolor flow cytometry experiments and is available exclusively from BD Biosciences. It is excited by the Violet laser Ex max of 406nm and has an Em Max at 450 nm. Conjugates with BD Horizon™ V450 can be used in place of Pacific Blue™ conjugates.
Specifications
| MIP-1b | |
| Monoclonal | |
| Violet Fluor 450 (V450) | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Recombinant Human MIP-1β | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| D21-1351 | |
| Macrophage inflammatory protein 1-beta; CCL4; C-C motif chemokine 4; LAG-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.