Learn More
MIP-1b Mouse anti-Human, PE, Clone: D21-1351, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561120
Description
The D21-1351 monoclonal antibody specifically binds to the human CC chemokine, MIP-1 β (macrophage inflammatory protein-1 β ). Human MIP-1 β shares approximately 75% homology with mouse MIP-1 β at the amino acid level. Expression of MIP-1 β in human peripheral blood cells is induced by proinflammatory and mitogenic stimuli. MIP-1 β is a chemoattractant for monocytes and lymphocytes. Human MIP-1 β
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Phamingen during antibody development only or reported in the literature.
Specifications
| MIP-1b | |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human MIP-1β | |
| 25 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| D21-1351 | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.