Learn More
MIP-1β Mouse anti-Human, Brilliant Violet 421, Clone: D21-1351, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562900
Description
The D21-1351 monoclonal antibody specifically binds to the human CC chemokine, MIP-1 β (macrophage inflammatory protein-1 β ). Human MIP-1 β shares approximately 75% homology with mouse MIP-1 β at the amino acid level. Expression of MIP-1 β in human peripheral blood cells is induced by proinflammatory and mitogenic stimuli. MIP-1 β is a chemoattractant for monocytes and lymphocytes. Human MIP-1 β
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon™ BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon™ BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Specifications
| MIP-1β | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Recombinant Human MIP-1β | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| D21-1351 | |
| Macrophage inflammatory protein 1-beta; CCL4; C-C motif chemokine 4; LAG-1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.