Learn More
MIG Mouse anti-Human, Biotin, Clone: B8-6, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555037
Description
The monoclonal antibody B8-6 reacts with human monokine induced by gamma interferon (MIG). MIG is inducible in macrophages, hepatocytes, and endothelial cells by IFN-γ, but not by IFN-α or bacterial lipopolysaccharides. The immunogen used to generate the monoclonal antibody B8-6 was insect cell-expressed, recombinant human MIG protein.
ELISA
Specifications
| MIG | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human MIG Protein | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
| ELISA | |
| B8-6 | |
| Biotin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.