Learn More
MIC A/B Mouse anti-Human, PE, Clone: 6D4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 558352
Description
The 6D4 monoclonal antibody specifically binds to the human MHC class I polypeptide-related sequence A (MICA, aka PERB11.1) and B (MICB. aka PERB11.2) proteins. These ∽70 kDa transmembrane glycoproteins are homologs of the major histocompatibility complex class I molecules although they lack association with β2 microglobulin. The MHC class I-related MICA and MICB chains are expressed by some gut epithelial cells in vivo. MICA and MICB expression by other epithelial cells and cell types, including fibroblasts and endothelial cells, is induced by stress, eg, stress caused by bacterial and viral infections, autoimmunity or cellular transformation. Epithelial cell expression of MICA and MICB has also been detected in transplanted kidneys and pancreas that show histological signs of rejection and or cellular injury. This suggests their potential role in transplant immunopathology. MICA and MICB are ligands for NKG2D (CD314), an activating receptor expressed by natural killer (NK) cells, γδ T cells, CD8+ and some CD4+ αβ T cells
Specifications
| MIC A/B | |
| Monoclonal | |
| PE | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| 6D4 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| MICA transfected cells | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.