missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HLA-DR/DP Monoclonal Antibody (HL-40)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA119147
This item is not returnable.
View return policy
Description
This antibody reacts with an extracellular common epitope on beta-chain of human HLA-DR and HLA-DP. DR and DP are the isotypes of human MHC Class II molecules expressed on antigen-presenting cells (APC; dendritic cells, B lymphocytes, monocytes, macrophages).
HLA-DRA is one of the HLA class II alpha chain paralogues. This class II molecule is a heterodimer consisting of an alpha and a beta chain, both anchored in the membrane. It plays a central role in the immune system by presenting peptides derived from extracellular proteins. Class II molecules are expressed in antigen presenting cells (APC: B lymphocytes, dendritic cells, macrophages). The alpha chain is approximately 33-35 kDa and its gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. [provided by RefSeq, Jul 2008]. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| HLA-DR/DP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| P01911, P04440 | |
| HLA-DPB1, HLA-DRB1 | |
| Burkitt's lymphoma cell line Raji. | |
| 100 μg | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
| HL-40 | |
| Unconjugated | |
| HLA-DPB1 | |
| beta1 domain MHC class II HLA DPB; class II HLA beta chain; Clone P2-beta-3; DPB1; DR-16; DRB1; DRw10; DW2.2/DR2.2; HLA class II histocompatibility antigen, DP beta 1 chain; HLA class II histocompatibility antigen, DP(W4) beta chain; HLA class II histocompatibility antigen, DR-1 beta chain; HLA class II histocompatibility antigen, DRB1-15 beta chain; HLA class II histocompatibility antigen, DRB1-16 beta chain; HLA class II histocompatibility antigen, DRB1-3 chain; HLA DP14-beta chain; HLA-D; HLA-DP; HLA-DP histocompatibility type, beta-1 subunit; HLA-DP1B; HLA-DPB; HLA-DPB1; HLA-DR; HLA-DR1B; HLA-DRB; HLA-DRB1; HLA-DRB2; human leucocyte antigen DRB1; lymphocyte antigen DRB1; major histocompatibility complex class II antigen beta chain; major histocompatibility complex, class II, DP beta 1; major histocompatibility complex, class II, DR beta 1; MHC Class 2; MHC class II; MHC class II antigen; MHC class II antigen beta chain; MHC class II antigen DP beta 1 chain; MHC class II antigen DPB1; MHC class II antigen DPbeta1; MHC class II antigen DRB1*15; MHC class II antigen DRB1*16; MHC class II antigen DRB1*3; MHC class II HLA-DP-beta-1; MHC class II HLA-DR beta 1 chain; MHC class II HLA-DRB1; MHC class II HLA-DR-beta cell surface glycoprotein; MHC class II HLA-DRw10-beta; MHC HLA DPB1; MHC HLA-D; MHC II HLA; SS1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3115, 3123 | |
| 4°C, do not freeze | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction