Learn More
HLA-ABC Monoclonal Antibody (W6/32), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA180453
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody does not react with amphibia, chicken, dog, goat, guinea pig, mouse or rabbit. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human HLA ABC antibody, clone W6/32 recognizes an antigenic determinant shared among products of the HLA A, B and C loci. Clone W6/32 recognizes a conformational epitope, reacting with HLA class I alpha3 and alpha2 domains.
HLA-class I major histocompatibility (MHC) antigens are intrinsic membrane glycoproteins expressed on nucleated cells and noncovalently associated with an invariant beta2 microglobulin. They carry foreign determinants important for immune recognition by cytotoxic T cells, thus important for anti-viral and anti-tumor defense. Human HLA-class I antigens are represented by HLA-A, HLA-B and HLA-C molecules. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.Specifications
HLA-ABC | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2 | |
P01889, P04439, P10321 | |
HLA-A, HLA-B, HLA-C | |
Purified human tonsil lymphocyte membranes. | |
200 μL | |
Primary | |
Bovine, Human, Non-human Primate, Shrew | |
Antibody | |
IgG2a |
Flow Cytometry | |
W6/32 | |
Biotin | |
HLA-C | |
A29 protein; alpha chain of MHC class II; antigen presenting molecule; AS; Aw-33; Aw-74; B-4901; B-5001; Bw-47; Bw-50; class I MHC, HLA-B antigen; D6S204; epididymis secretory protein Li 83; H-2K; H2-K; H-2K(d); HEL-S-83; histocompatibility antigen; HLA A; HLA -B; HLA B-40011; HLA B44; HLA C; HLA class I antigen; HLA class I antigen HLA-B; HLA class I histocompatibility antigen C alpha chain; HLA class I histocompatibility antigen Cw-1 alpha chain; HLA class I histocompatibility antigen, A alpha chain; HLA class I histocompatibility antigen, A-1 alpha chain; HLA class I histocompatibility antigen, A-11 alpha chain-like; HLA class I histocompatibility antigen, A-30 alpha chain; HLA class I histocompatibility antigen, A-33 alpha chain; HLA class I histocompatibility antigen, A-74 alpha chain; HLA class I histocompatibility antigen, B alpha chain; HLA class I histocompatibility antigen, B-47 alpha chain; HLA class I histocompatibility antigen, B-50 alpha chain; HLA class I histocompatibil | |
Mouse | |
Affinity chromatography | |
RUO | |
3105, 3106, 3107 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |