missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mGluR2/mGluR3 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA536519

Product Code. 15503911

  • £441.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody detects endogenous protein at a molecular weight of 95 kDa. Purity is >95% by SDS-PAGE.

The mGluR proteins (metabotropic Glutamate Receptors) are members of the G-protein-coupled receptor family and are functionally and pharmacologically distinct from the GluR proteins (ionotropic Glutamate Receptors). The eight currently known mGluR proteins are mediated by two G-proteins with opposing regulation of adenylate cyclase pathways. The activities of mGluR1 and mGluR5 are mediated by a G-protein that activates a phosphatidylinositol-calcium second messenger system and generates a calcium-activated chloride current. The remainder of the eight sub-types of mGluR have an activity mediated by a G-protein that inhibits adenylate cyclase activity. mGluR-3, which may interact with GRASP, acts as a receptor for glutamate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

mGluR2/mGluR3
Polyclonal
Unconjugated
Grm2
0710001G23Rik; 4930441L02Rik; G protein coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member B; G protein-coupled receptor, family C, group 1, member C; GLUR2; GLUR3; glutamate metabotropic receptor 2; glutamate metabotropic receptor 3; glutamate receptor; glutamate receptor homolog; glutamate receptor, metabotropic 2; glutamate receptor, metabotropic 3; Gprc1b; GPRC1C; GRM2; Grm3; M mGlu3; Metabotropic glutamate receptor 2; metabotropic glutamate receptor 3; mGlu2; mGlu3; mGluR2; MGLUR3; mGluR7
Rabbit
Antigen Affinity Chromatography
RUO
108068, 108069, 24415, 24416, 2912, 2913
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
P31421, P31422, Q14416, Q14832, Q14BI2, Q9QYS2
Grm2, Grm3
Synthetic peptide corresponding to amino acids 836-872 of Human mGluR-2.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.