Learn More
MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant MFNG.
Brand: Abnova H00004242-A01.50uL
Description
This gene is a member of the fringe gene family which also includes Radical and Lunatic fringe. They all encode evolutionarily conserved secreted proteins that act in the Notch receptor pathway to demarcate boundaries during embryonic development. While their genomic structure is distinct from other glycosyltransferases, fringe proteins have a fucose-specific beta1,3 N-acetylglucosaminyltransferase activity that leads to elongation of O-linked fucose residues on Notch, which alters Notch signaling. [provided by RefSeq
Sequence: WASGSRFMDTSALIRLPDDCTMGYIIECKLGGRLQPSPLFHSHLETLQLLRTAQLPEQVTLSYGVFEGKLNVIKLQGPSpecifications
| MFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant MFNG. | |
| MFNG | |
| MFNG | |
| MFNG (NP_002396, 214 a.a. ∼ 291 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_002405 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 4242 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.