Learn More
mesoderm posterior 2 homolog (mouse), Mouse, Clone: 1C8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MESP2.
Brand: Abnova H00145873-M01.100ug
Description
This gene encodes a member of the bHLH family of transcription factors and plays a key role in defining the rostrocaudal patterning of somites via interactions with multiple Notch signaling pathways. This gene is expressed in the anterior presomitic mesoderm and is downregulated immediately after the formation of segmented somites. This gene also plays a role in the formation of epithelial somitic mesoderm and cardiac mesoderm. Mutations in the MESP2 gene cause autosomal recessive spondylocostal dystosis 2 (SCD02). [provided by RefSeq
Sequence: AQSPPPQSLLGHDHWIFAQGWGWAGHWDSTSPASSSDSSGSCPCDGARGLPQPQPPSCSSRAAEAAATTPRRARTGPAGGQRQSASEREKLRMRTLARALHELRRFLPPSLAPAGQSLTKIETLSpecifications
| mesoderm posterior 2 homolog (mouse) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| XM_085261 | |
| MESP2 | |
| MESP2 (XP_085261, 2 a.a. ∼ 125 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence | |
| 1C8 | |
| Mouse monoclonal antibody raised against a partial recombinant MESP2. | |
| MESP2 | |
| SCDO2/bHLHc6 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 145873 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.