missing translation for 'onlineSavingsMsg'
Learn More

Mena Mouse, Unlabeled, Clone: 21, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610693

 View more versions of this product

Product Code. 15865589

  • ONLINE EXCLUSIVE PRICE
    £303.00
    £290.95 / 1 set
    Save £12.05
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Abl and Disabled (Dab) are essential for correct axonal connections during Drosophila development. Mutations in Enabled (Ena) rescue the genetic defects caused by mutated Abl and Dab. Mena (Mammalian Ena) is a protein with significant homology to Eva, VASP, and WASP proteins. The murine Mena gene predicts a protein of 541 amino acids. However, two additional exons that could introduce 246 amino acids onto the C-terminal region have been identified. Homology searches identified two EVH domains (Ena-Vasp homology), one near each terminus of Mena. Antibodies to Mena detect 80 kDa, 88 kDa, and 140 kDa protein with the 80 kDa and 88 kDa proteins being widely expressed and the 140 kDa protein reportedly mainly found in brain tissue. Like VASP, Mena is localized to focal contacts and, when ectopically expressed, induces the formation of F-actin outgrowths in fibroblasts.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mena
Monoclonal
250μg/mL
Mammalian Ena (Enabled)
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
21
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
150 μg
Cell Biology
Chicken, Human, Mouse, Rat
IgA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.