missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDM1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539310
Code : NEW
Additional Details : Weight : 0.00900kg
Description
This gene encodes a nuclear protein similar to the mouse double minute 1 protein. The mouse gene is located in double minute (DM) chromatin particles, is amplified in the mouse transformed 3T3 cell line, and the encoded protein is able to bind to p53. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Mar 2011].Specifications
MDM1 | |
Polyclonal | |
Unconjugated | |
MDM1 | |
MDM1 | |
A synthetic peptide derived from the C-terminal region of human MDM1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8TC05, Q9D067, Q5PQN4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17245, 314859, 56890 | |
-20°C | |
Liquid |