missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NG2 Monoclonal Antibody (LHM 2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA190928
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA1-90928 detects MCSP from human samples.
NG2 Monoclonal antibody specifically detects NG2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.Specifications
NG2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q6UVK1 | |
Cspg4 | |
Tissue / cell preparation (Human - a 375P cell crude extract). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
LHM 2 | |
Unconjugated | |
Cspg4 | |
4732461B14Rik; AN2; AN2 proteoglycan; AN2/NG2 proteoglycan; chondroitin sulfate proteoglycan 4; chondroitin sulfate proteoglycan 4 (melanoma-associated); chondroitin sulfate proteoglycan NG2; CSPG4; HMW MAA; HMW-MAA; HSN tumor-specific antigen; Kiaa4232; MCSP; MCSPG; MEL CSPG; melanoma chondroitin sulfate proteoglycan; Melanoma-associated chondroitin sulfate proteoglycan; MEL-CSPG; membrane-spanning proteoglycan NG2; MSK16; nerve/glial antigen 2; Ng2; Proteoglycan AN2 | |
Mouse | |
IgG fraction | |
RUO | |
1464 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |