missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCP-5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132933
Code : LE
Additional Details : Weight : 0.01000kg
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Chemotactic factor that attracts eosinophils, monocytes, and lymphocytes but not neutrophils. Potent monocyte active chemokine that signals through CCR2. Involved in allergic inflammation and the host response to pathogens and may play a pivotal role during early stages of allergic lung inflammation.Specifications
MCP-5 | |
Polyclonal | |
Unconjugated | |
Ccl12 | |
C-C motif chemokine 12; Ccl12; chemokine (C-C motif) ligand 12; MCP-1-related chemokine; Mcp5; MCP-5; Monocyte chemoattractant protein 5; monocyte chemotactic protein 5; Scya12; small inducible cytokine A12; small-inducible cytokine A12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
20293 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Functional Assay, Western Blot | |
1 mg/mL | |
PBS with no preservative | |
Q62401 | |
Ccl12 | |
Recombinant mouse MCP-5 | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |