Learn More
MCP-1 Hamster anti-Mouse, NA/LE, Unlabeled, Clone: 2H5, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 554440
Description
The 2H5 antibody reacts with mouse and rat monocyte chemoattractant protein (MCP-1), formerly termed JE. This antibody also recognizes human MCP-1, but shows no reactivity with the closely related mouse β chemokines, TCA3 and MIP-1β. The immunogen used to generate the 2H5 hybridoma was heparin-purified CHO-expressed mouse MCP-1. This is a neutralizing antibody.
Specifications
| MCP-1 | |
| Monoclonal | |
| 1.0mg/mL | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| Heparin-purified CHO-expressed mouse MCP-1 | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
| ELISA | |
| 2H5 | |
| Unconjugated | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.