missing translation for 'onlineSavingsMsg'
Learn More

MCM6 Mouse anti-Human, Unlabeled, Clone: 1, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611622

Product Code. 15815999

  • ONLINE EXCLUSIVE PRICE
    £171.00
    £163.84 / 1 set
    Save £7.16
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Rabbit anti-Goat IgG (H+L) Secondary Antibody, AP conjugate, 61-1622, from Invitrogen™. Species Reactivity: Goat; Applications: ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Host Species: Rabbit
Cross Reactivity: Goat
Isotype: IgG
Immunogen: Gamma Immunoglobin
Formulation: 1mL purified antibody in 50mM tris with 1% BSA, 40% glycerol and 0.1% sodium azide
This product detects Goat IgG (H+L) in Goat samples. This product has been successfully used in ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation and Western Blot procedures. The immunogen is Gamma Immunoglobin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MCM6
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human MCM6 aa. 670-792
50 μg
Cell Biology
Human
IgG1
Western Blot
1
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.