Learn More
malonyl CoA:ACP acyltransferase (mitochondrial), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human MCAT protein.
Brand: Abnova H00027349-D01P.100ug
Description
The protein encoded by this gene is found exclusively in the mitochondrion, where it catalyzes the transfer of a malonyl group from malonyl-CoA to the mitochondrial acyl carrier protein. The encoded protein may be part of a fatty acid synthase complex that is more like the type II prokaryotic and plastid complexes rather than the type I human cytosolic complex. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MSVRVARVAWVRGLGASYRRGASSFPVPPPGAQGVAELLRDATGAEEEAPWAATERRMPGQCSVLLFPGQGSQVVGMGRGLLNYPRVRELYAAARRVLGYDLLELSLHGPQETLDRTVHCQPAIFVASLAAVEKLHHLQPSVIENCVAAAGFSVGEFAALVFAGAMEFAEGSTVSPEEFLSpecifications
| malonyl CoA:ACP acyltransferase (mitochondrial) | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human MCAT protein. | |
| MCAT | |
| FASN2C/MCT/MGC47838/MT/fabD | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 27349 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014507.2 | |
| MCAT | |
| MCAT (NP_055322.1, 1 a.a. ∼ 180 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.