missing translation for 'onlineSavingsMsg'
Learn More
Learn More
muscleblind-like (Drosophila), Mouse, Clone: 1D11, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant MBNL1.
Brand: Abnova H00004154-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: MGRCSRENCKYLHPPPHLKTQLEINGRNNLIQQKNMAMLAQQMQLANAMMPGAPLQPVPMFSVAPSLATNASAAAFNPYLGPVSPSLVPAEILPTAPMLVTGNPGVPVPAAAAAAAQKLMRTDRLEVCREYQRGNCNRGENDCRFAHPADSTMIDTNDNTVTVCMDYIKGRCSREKCKYFHPPAHLQAKIKAAQYQVNQAAAAQAAATAAAMTQSAVKSLKRPLEATFDLGIPQAVLPPLPKRPALEKTNGATAVFNTGIFQYQQALANMQLQQHTAFLPPGSILCMTPATSVVPMVHGATPATVSAATTSATSVPFAATATANQIPIISAEHLTSHKYVTQMSpecifications
| muscleblind-like (Drosophila) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC050535 | |
| MBNL1 | |
| MBNL1 (AAH50535, 1 a.a. ∼ 343 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 4154 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 1D11 | |
| Mouse monoclonal antibody raised against a full length recombinant MBNL1. | |
| MBNL1 | |
| DKFZp686P06174/EXP/EXP35/EXP40/EXP42/KIAA0428/MBNL | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction