missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enolase-phosphatase 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MASA protein.
Brand: Abnova H00058478-B01.50uL
This item is not returnable.
View return policy
Description
Sequence: MVVLSVPAEVTVILLDIEGTTTPIAFVKDILFPYIEENVKEYLQTHWEEEECQQDVSLLRKQAEEDAHLDGAVPIPAASGNGVDDLQQMIQAVVDNVCWQMSLDRKTTALKQLQGHMWRAAFTAGRMKAEFFADVVPAVRKWREAGMKVYIYSSGSVEAQKLLFGHSTEGDILELVDGHFDTKIGHKVESESYRKIADSIGCSTNNILFLTDVTREASAAEEADVHVAVVVRPGNAGLTDDEKTYYSLITSFSELYLPSSTSpecifications
| enolase-phosphatase 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MASA protein. | |
| ENOPH1 | |
| DKFZp586M0524/E1/FLJ12594/MASA/MST145 | |
| Mouse | |
| 50 μL | |
| Stem Cell Biology | |
| Primary | |
| Human | |
| Serum |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_021204 | |
| ENOPH1 | |
| MASA (NP_067027, 1 a.a. ∼ 261 a.a) full-length human protein. | |
| RUO | |
| Yes | |
| 58478 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction