Learn More
MARCO, PE, clone: ED31, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516662
Description
The protein encoded by this gene is a member of the class A scavenger receptor family and is part of the innate antimicrobial immune system. The protein may bind both Gram-negative and Gram-positive bacteria via an extracellular, C-terminal, scavenger receptor cysteine-rich (SRCR) domain. In addition to short cytoplasmic and transmembrane domains, there is an extracellular spacer domain and a long, extracellular collagenous domain. The protein may form a trimeric molecule by the association of the collagenous domains of three identical polypeptide chains.
Reconstitute with 1 ml of distilled water. After reconstitution, store product undiluted at 4ºC in the dark.For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.
Specifications
| MARCO | |
| Monoclonal | |
| PE | |
| Q60754 | |
| MARCO | |
| CHO cells expressing mouse MARCO | |
| 100 Tests | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| ED31 | |
| Marco | |
| AI323439, Ly112, Scara2,macrophage receptor MARCO | |
| Rat | |
| Protein G | |
| RUO | |
| 17167 | |
| 4° C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.