Learn More
MAPKAPK-2 Mouse, PE, Clone: P24-694, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562470
Description
The P24-694 monoclonal antibody specifically binds to the phosphorylated T334 site (pT334) of MAPKAPK-2. MAPKAPK-2 is a serine/threonine protein kinase. This ∽49 kDa member of the MAPKAPK family of protein kinases is also known as mitogen-activated protein kinase-activated protein kinase 2. MAPKAPK-2 is phosphorylated and activated by p38 MAP kinase in response to stress, cytokines and chemokines. MAPKAPK-2 is phosphorylated on multiple sites including Thr222, Ser272 and Thr334. Phosphorylation of any two of these three amino acid residues seems to be required for the activation of this kinase thatserves multiple cellular functions. Phosphorylation of Thr334 was reported to be essential for nuclear export of the heterodimer formed between p38 MAPK and MAPKAPK-2. Mice deficient in MAPKAPK-2 have been shown to be protected from ischemic injury. MAPKAPK-2 is also reported to serve as a cell cycle checkpoint kinase in response to UV irradiation. The heat shock protein, HSP27 was shown to be one of the major substrates of MAPK and MAPKAPK-2.
Specifications
| MAPKAPK-2 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Phosphorylated human MAPKAPK-2 peptide | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| P24-694 | |
| MAP kinase-activated protein kinase 2, MAPKAP kinase 2; MK2; MAPK2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.