Learn More
mitogen-activated protein kinase kinase kinase kinase 4, Mouse, Clone: 4A5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MAP4K4.
Brand: Abnova H00009448-M07.100ug
Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. This kinase has been shown to specifically activate MAPK8/JNK. The activation of MAPK8 by this kinase is found to be inhibited by the dominant-negative mutants of MAP3K7/TAK1, MAP2K4/MKK4, and MAP2K7/MKK7, which suggests that this kinase may function through the MAP3K7-MAP2K4-MAP2K7 kinase cascade, and mediate the TNF-alpha signaling pathway. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq
Sequence: VHPALQRPAEPQVQWSHLASLKNNVSPVSRSHSFSDPSPKFAHHHLRSQDPCPPSRSEVLSQSSDSKSEAPDPTQKAWSRSDSDEVPPRVPVRTTSRSPVSpecifications
| mitogen-activated protein kinase kinase kinase kinase 4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_145686 | |
| MAP4K4 | |
| MAP4K4 (NP_663719, 611 a.a. ∼ 710 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 4A5 | |
| Mouse monoclonal antibody raised against a partial recombinant MAP4K4. | |
| MAP4K4 | |
| FLH21957/FLJ10410/FLJ20373/FLJ90111/HGK/KIAA0687/NIK | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 9448 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.