missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-MAP3K5, ERN1 Protein Protein Interaction Antibody Pair, Abnova™
Brand: Abnova DI0169.1Set
Includes: MAP3K5 rabbit purified polyclonal antibody (20μg); ERN1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Description
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAP3K5 protein, and the other against the ERN1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Specifications
| MAP3K5 | |
| Monoclonal/Polyclonal | |
| MAP3K5 | |
| MAP3K5 | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. MAP3K5 rabbit purified polyclonal antibody (20 ug) 2. ERN1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
| Purified |
| In situ PLA | |
| Unconjugated | |
| ASK1/MAPKKK5/MEKK5 | |
| Mouse, Rabbit | |
| RUO | |
| 4217/2081 | |
| Human | |
| MAP3K5 & ERN1 Protein Protein Interaction Antibody Pair |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction