Learn More
MAP2B Mouse, Unlabeled, Clone: 18/MAP2B, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610460
Description
Microtubule-associated proteins (MAPs) play a crucial role in the development and morphology of neurons. MAP2, specifically localized in dendrites, has four known isoforms that are produced by alternative splicing of the transcript and are expressed at various stages of neuronal development. MAP2B is a 280-kDa protein that is expressed throughout brain development. It contains functional domains that interact with the regulatory subunit of the cAMP-dependent kinase II and microtubules. Experimental monitoring of its presence, along with GFAP and nestin, may be utilized to quantify neuronal development.
Specifications
| MAP2B | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG1 κ |
| Western Blot | |
| 18/MAP2B | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.