Learn More
anti-MALT1 Polyclonal, Invitrogen™
MALT1 Polyclonal antibody A16538 from Thermo Scientific™ Pierce™ specifically detects MALT1 in Human samples, and it is validated for WB and IP
Brand: Invitrogen A16538
Description
MALT1/Paracaspase (MALT1) was independently identified as a member of the human paracaspase family and an interacting partner of B-cell lymphoma (Uren et al., 2000). MALT1 is a caspase-like protein that contains an N-terminal death domain, two Ig-like domains, and a C-terminal caspase-like domain. It binds to Bcl10 through its Ig-like domains and cooperates with Bcl10 to activate NF-kappaB.
A16538 detects MALT1 in Human samples. A16538 has been successfully used in WB and IP procedures. The immunogen for A16538 is a synthetic peptide corresponding to the carboxy terminus of mouse and rat MALT1 protein.
Applications:
Immunoprecipitation, Western Blotting
Specifications
| MALT1 | |
| Unconjugated | |
| Rabbit | |
| Primary | |
| Human | |
| Purified |
| Polyclonal | |
| Q9UDY8 | |
| A synthetic peptide corresponding to the carboxy terminus of mouse and rat MALT1 protein | |
| 10892 | |
| Store at -20°C | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.