Learn More
melanoma antigen family D, 1 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human MAGED1 protein.
Brand: Abnova H00009500-B01.50uL
Description
This gene is a member of the melanoma antigen gene (MAGE) family. Most of the genes of this family encode tumor specific antigens that are not expressed in normal adult tissues except testis. Although the protein encoded by this gene shares strong homology with members of the MAGE family, it is expressed in almost all normal adult tissues. This gene has been demonstrated to be involved in the p75 neurotrophin receptor mediated programmed cell death pathway. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq
Sequence: MAQKMDCGAGLLGFQAEASVEDSALLMQTLMEAIQISEAPPTNQATAAASPQSSQPPTANEMADIQVSAAAARPKSAFKVQNATTKGPNGVYDFSQAHNAKDVPNTQPKAAFKSQNATPKGPNAAYDFSQAATTGELAANKSEMAFKAQNATTKVGPNATYNFSQSLNANDLANSRPKTPFKAWNDTTKAPTADTQTQNVNQAKMATSQADIETDPGISEPDGATAQTSADGSQAQNLESRTIIRGKRTRKINNLNVEENSSGDQRRAPLAAGTWRSAPVPVTTQNPPGAPPNVLWQTPLAWQNPSGWQNQTARQTPPARQSPPARQTPPAWQNPVAWQNPVIWPNPVIWQNPVIWPNPIVWPGPVVWPNPLAWQNPPGWQTPPGWQTPPGWQGPPDWQGPPDWPLPPDWPLPPDWPLPTDWPLPPDWIPADWPIPPDWQNLRPSPNLRPSPNSRASQNPGAAQPRDVALLQERANKLVKYLMLKDYTKVPIKRSEMLRDIIREYTDVYPEIIERACFVLEKKFGIQLKEIDKEEHLYILISTPESLAGILGTTKDTPKLGLLLVILGVIFMNGNRASEAVLWEALRKMGLRPGVRHPLLGDLRKLLTYEFVKQKYLDYRRVPNSNPPEYEFLWGLRSYHETSKMKVLRFIAEVQKRDPRDWTAQFMEAADEALDALDAAAAEAEARAEARTRMGIGDEAVSGPWSWDDIEFELLTWDEEGDFGDPWSRIPFTFWARYHQNARSRFPQTFAGPIIGPGGTASANFAANFGAIGFFWVESpecifications
| melanoma antigen family D, 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human MAGED1 protein. | |
| MAGED1 | |
| DLXIN-1/NRAGE | |
| Mouse | |
| 50 μL | |
| Yes | |
| 9500 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| NM_001005332.1 | |
| MAGED1 | |
| MAGED1 (NP_001005332.1, 1 a.a. ∼ 778 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.