Learn More
MAD2 Mouse, Unlabeled, Clone: 48, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610678
Description
Progression of the mammalian cell cycle is regulated by phosphorylation/dephosphorylation and synthesis/degradation of many key proteins. These events are of utmost importance at the checkpoints, or transition points, of the cell cycle. MAD2 (Mitotic Arrest Deficient) is the human homolog of a yeast and Xenopus protein that is essential for spindle assembly during mitosis. The human hsMAD2 gene encodes a protein of 205 amino acids with a predicted molecular weight of 23.5 kDa. Binding of affinity purified polyclonal antibodies to the MAD2 protein prevents mitosis of HeLa cells. This indicates that, like its invertebrate relatives, MAD2 is necessary for mitosis. Furthermore, MAD2 is localized at the kinetochore of condensed chromosomes during mitosis and cells defective in the mitotic checkpoint have reduced levels of MAD2.
Specifications
| MAD2 | |
| Monoclonal | |
| 250μg/mL | |
| Mitotic Arrest Deficient-2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 48 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Human, Mouse, Rat | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.