Learn More
M-cadherin Mouse anti-Human, Clone: 21G4, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA191683
Description
MA1-91683 detects M Cadherin from human samples. MA1-91683 has been successfully used in immunofluorescence, Western blot applications. The MA1-91683 immunogen is: Recombit full length protein - the extracellular domain of human M-Cadherin Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
This gene encodes a type I integral membrane glycoprotein. The encoded protein acts as a receptor and binds to both soluble and immobilized hyaluro. This protein may function in lymphatic hyaluro transport and have a role in tumor metastasis.
Specifications
| M-cadherin | |
| Monoclonal | |
| 2.0 mg/mL | |
| CDH15 | |
| CRSBP-1, HAR, LYVE-1, XLKD1 | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 1013 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 21G4 | |
| Unconjugated | |
| P55291 | |
| CDH15 | |
| Recombit full length protein - the extracellular domain of human M-Cadherin | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.