Learn More
LZTS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA544945
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: EPLCPAVPPR KAVPVTSFTY INEDFRTESP PSPSSDVEDA REQRAHNAHL Sequence homology: Cow: 100%; Dog: 100%; Goat: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%
LZTS2 Polyclonal antibody specifically detects LZTS2 in Human samples. It is validated for Western Blot.Specifications
LZTS2 | |
Polyclonal | |
Unconjugated | |
LZTS2 | |
BC014695; HAMAP-Rule:MF_03026}; hLZTS2; KIAA1813; LAPSER1; Leucine zipper putative tumor suppressor 2; leucine zipper putative tumor suppressor 2 {ECO:0000255; leucine zipper tumor suppressor 2; leucine zipper, putative tumor suppressor 2; LOW QUALITY PROTEIN: leucine zipper putative tumor suppressor 2; Lzts2; mKIAA1813; Protein LAPSER1; putative tumor suppressor LAPSER1; RP11-108L7.8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
84445 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BRK4 | |
LZTS2 | |
Synthetic peptide directed towards the N-terminal of human LZTS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |