missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Luteinizing Hormone Mouse anti-Human, Clone: LH01, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512138
This item is not returnable.
View return policy
Description
MA5-12138 targets Luteinizing Hormone in IHC (P) applications and shows reactivity with Human samples. The MA5-12138 immunogen is purified human LH-beta.
Luteinizing hormone (LH) is a tropic hormone which modulates the secretory activity of other endocrine glands. It is produced in the anterior hypophysis of the pituitary gland. The glycoprotein hormone, LH, like follicle stimulating hormone and thyroid stimulating hormone, is composed of a common alpha-subunit but also a spesofic beta-subunit, which characterizes each of these hormones.
Specifications
| Luteinizing Hormone | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| P01229, P01215 | |
| CGA, LHB | |
| Purified human LH-beta | |
| 500 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| LH01 | |
| Unconjugated | |
| CGA | |
| LSH-beta, interstitial cell stimulating hormone, beta chain, luteinizing hormone beta subunit, lutropin beta chain, lutropin subunit beta | |
| Mouse | |
| Protein G | |
| RUO | |
| 1081, 3972 | |
| 4° C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction