missing translation for 'onlineSavingsMsg'
Learn More

anti-Luciferase, Biotin, Polyclonal, Novus Biologicals™

Product Code. 18394192 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18394192 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18394192 Supplier Novus Biologicals Supplier No. NBP311675JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NALP6 Monoclonal specifically detects NALP6 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Frozen.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Luciferase
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation 0.02M Potassium phosphate, 0.15M NaCl (pH 7.2) with 10mg/mL BSA
Host Species Goat
Immunogen Luciferase [Photinus pyralis (Firefly)].
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Luciferase. Cross-reacts with Firefly. Not yet tested in other species.
Target Species Firefly
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.