missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lucifer Yellow Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen A5750
Code : 50
Additional Details : Weight : 0.00450kg
Description
Lucifer Yellow Polyclonal antibody specifically detects Lucifer Yellow in Chemical samples. It is validated for Immunocytochemistry, Immunohistochemistry.Specifications
Lucifer Yellow | |
Polyclonal | |
Unconjugated | |
Lucifer Yellow Dye | |
Lucifer Yellow | |
500 μL | |
Primary | |
Antibody | |
IgG |
Immunocytochemistry, Immunohistochemistry | |
3 mg/mL | |
PBS with 5mM sodium azide; pH 7.2 | |
Rabbit | |
IgG fraction | |
RUO | |
4° C | |
Liquid | |
Chemical |