Learn More
LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae), Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant LSM2.
Brand: Abnova H00057819-A01.50uL
Description
Sm-like proteins were identified in a variety of organisms based on sequence homology with the Sm protein family (see SNRPD2; MIM 601061). Sm-like proteins contain the Sm sequence motif, which consists of 2 regions separated by a linker of variable length that folds as a loop. The Sm-like proteins are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing.[supplied by OMIM
Sequence: MLFYSFFKSLVGKDVVVELKNDLSICGTLHSVDQYLNIKLTDISVTDPEKYPHMLSVKNCFIRGSVVRYVQLPADEVDTQLLQDAARKEALQQKQSpecifications
| LSM2 homolog, U6 small nuclear RNA associated (S. cerevisiae) | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant LSM2. | |
| LSM2 | |
| C6orf28/G7b/YBL026W/snRNP | |
| Mouse | |
| 50 μL | |
| Yes | |
| 57819 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_021177 | |
| LSM2 | |
| LSM2 (NP_067000, 1 a.a. ∼ 95 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.