Learn More
LSF Mouse, Unlabeled, Clone: 14, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610818
Description
LSF (αCP2 or LBP-1c) is a transcription factor originally identified by its binding to, and induction of, the SV40 late promoter. LSF displays some similarity to the Elf-1/NTF transcription factors of Drosophila. It is primarily found in
Immunofluorescence, Western Blotting
Specifications
| LSF | |
| Monoclonal | |
| 250μg/mL | |
| αCP2; LBP-1c | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 14 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human LSF aa. 205-414 | |
| 50 μg | |
| Cell Biology | |
| Canine, Human, Mouse, Rat | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.