Learn More
low density lipoprotein receptor-related protein 8, apolipoprotein e receptor, Mouse, Clone: 3H2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LRP8.
Brand: Abnova H00007804-M01.100ug
Description
This gene encodes an apolipoprotein E receptor, a member of the low density lipoprotein receptor (LDLR) family. Apolipoprotein E is a small lipophilic plasma protein and a component of lipoproteins such as chylomicron remnants, very low density lipoprotein (VLDL), and high density lipoprotein (HDL). The apolipoprotein E receptor is involved in cellular recognition and internalization of these lipoproteins. Alternative splicing generates multiple transcript variants encoding distinct isoforms for this gene. [provided by RefSeq
Sequence: KKTCADSDFTCDNGHCIHERWKCDGEEECPDGSDESEATCTKQVCPAEKLSCGPTSHKCVPASWRCDGEKDCEGGADEAGCATLCAPHSpecifications
| low density lipoprotein receptor-related protein 8, apolipoprotein e receptor | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004631 | |
| LRP8 | |
| LRP8 (NP_004622, 83 a.a. ∼ 170 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Membrane Receptors | |
| 7804 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Western Blot | |
| 3H2 | |
| Mouse monoclonal antibody raised against a partial recombinant LRP8. | |
| LRP8 | |
| APOER2/HSZ75190/MCI1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.