Learn More
LRP Mouse, Unlabeled, Clone: 42, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610512
Description
Drugs that selectively attack dividing cells are often used in cancer therapy. However, small cell populations do not die, but develop resistance to a variety of toxic drugs. Multidrug resistance is achieved by gene amplification of specific membrane-bound transport ATPases that shuttle the drugs out of cells. Some multidrug-resistant cancer cells express a protein of 110 kDa named LRP (Lung Resistance-related Protein). LRP expression might be an indicator of prognosis after chemotherapy in acute myeloid leukemia and ovarian carcinoma. The LRP gene encodes a protein of 896 amino acids with significant homology to the major vault protein from Dictyostelium discoideum. Although ubiquitously expressed, LRP is most abundant in epithelial cells.
Specifications
| LRP | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 50 μg | |
| Cell Biology | |
| Bovine, Canine, Human, Rabbit | |
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| 42 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.