Learn More
Invitrogen™ LOXL1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA544213
Product Code. 15526402
  • £443.00 / 100µL
Description
Peptide sequence: RWRQLIQWEN NGQVYSLLNS GSEYVPAGPQ RSESSSRVLL AGAPQAQQRR Sequence homology: Cow: 83%; Guinea Pig: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%.
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LOXL1
Polyclonal
Unconjugated
Loxl1
cross-linking enzyme; LOL; Lox2; LOXL; LOXL1; Lysyl oxidase 2; lysyl oxidase homolog 1; lysyl oxidase like 1; lysyl oxidase-like 1; lysyl oxidase-like protein 1
Rabbit
Affinity chromatography
RUO
4016
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q08397
Loxl1
Synthetic peptide directed towards the N-terminal of human LOXL1 (aa 38-87).