missing translation for 'onlineSavingsMsg'
Learn More

LOX Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA523001

Product Code. 13278078

  • £416.12 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The target sequence has 100% sequence homology with rat.

Lysyl oxidase (LOX) is an extracellular enzyme that is critical for the normal biosynthesis of collagens and elastin. LOX is synthesized as a 50 kDa precursor, secreted into the extracellular matrix and subsequently proteolytically processed to a 30 kDa active enzyme and an 18 kDa propeptide. LOX also reverts ras-mediated transformation and its expression is down-regulated in human cancers. Most recently, the lysyl oxidase pro-peptide (LOPP) has been associated with tumor suppression, as it inhibits ras-dependent cellular transformation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LOX
Polyclonal
Unconjugated
P28300, P28301
LOX
A synthetic peptide made to an internal region of mouse LOX propeptide (within residues 135-155).
100 μL
Primary
Human, Mouse
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
1 mg/mL
LOX
protein-lysine 6-oxidase
Rabbit
Antigen affinity chromatography
RUO
16948, 4015
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.