missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LONRF1 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA542356

Product Code. 15545412

  • £450.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Peptide sequence: MSSPAVARTS PGGSREMAPA PQGRGRFWEV GGGSGHRLER AAAESERWEL Sequence homology: Human: 100%; Mouse: 86%; Rat: 86%.

The function remains unknown.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LONRF1
Polyclonal
Unconjugated
LONRF1
LON peptidase N-terminal domain and ring finger 1; LON peptidase N-terminal domain and RING finger protein 1; LONRF1; RGD1562583; RING finger protein 191; RNF191
Rabbit
Protein A
RUO
91694
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
Q17RB8
LONRF1
Synthetic peptide directed towards the N-terminal of human LONRF1 (aa 1-50).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.