missing translation for 'onlineSavingsMsg'
Learn More
Learn More
proline rich 16, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant LOC51334.
Brand: Abnova H00051334-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: MTDSSKTDTLNSSSSGTTASSLEKIKVQANAPLIKPPAHTSAILTVLRKPNPPPPPPRLTPVKCEDPKRVVPTANPVKTNGTLLRNGGLPGGPNKIPNGDICCIPNSNLDKAPVQLLMHRPEKDRCPQAGPRERVRFNEKVQYHGYCPDCDTRYNIKNREVHLHSEPVHPPGKIPHQGPPLPPTPHLPPFPLENGGMGISHSNSFPPIRPATVPPPTAPKPQKTILRKSTTTTVSpecifications
| proline rich 16 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant LOC51334. | |
| PRR16 | |
| DSC54/MGC104614 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 51334 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC038838 | |
| PRR16 | |
| LOC51334 (AAH38838.1, 1 a.a. ∼ 234 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction