missing translation for 'onlineSavingsMsg'
Learn More

cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant CMPK2.

Brand:  Abnova H00129607-A01.50uL

Product Code. 16101767

  • ONLINE EXCLUSIVE PRICE
    £209.00
    £179.64 / 50µL
    Save £29.36
    14% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Mitochondrial UMP-CMP kinase (EC 2.7.2.14) is a component of the salvage pathway for nucleotide synthesis. Other enzymes of the salvage pathway include thymidine kinase-2 (TK2; MIM 188250), deoxynucleotidase-2 (NT5M; MIM 605292), deoxyguanosine kinase (DGUOK; MIM 601465), adenylate kinase-2 (AK2; MIM 103020), adenylate kinase-3 (AK3; MIM 609290), adenylate kinase-3-like-1 (AK3L1; MIM 103030), and nucleoside diphosphate kinase (NME4; MIM 601818) (Xu et al., 2008 [PubMed 17999954]).[supplied by OMIM

Sequence: GACQEAPRPHLGEFEADPRGQLWQRLWEVQDGRRLQVGCAQVVPVPEPPLHPVVPDLPSSVVFPDREAARAVLEECTSFIPEARAVLDLVDQCPKQIQKG
Specifications

Specifications

cytidine monophosphate (UMP-CMP) kinase 2, mitochondrial
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant CMPK2.
CMPK2
TYKi/UMP-CMPK2
Mouse
50 μL
Kinases
Primary
Human
Serum
ELISA, Western Blot
Unconjugated
50% glycerol
NM_207315
CMPK2
CMPK2 (NP_997198, 151 a.a. ∼ 250 a.a) partial recombinant protein with GST tag.
RUO
Yes
129607
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.