missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM domain only 4, Mouse, Clone: 3B4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant LMO4.
Brand: Abnova H00008543-M04.100ug
This item is not returnable.
View return policy
Description
This gene encodes a cysteine-rich protein that contains two LIM domains but lacks a DNA-binding homeodomain. The encoded protein may play a role as a transcriptional regulator or as an oncogene. [provided by RefSeq
Sequence: LGDIGTSCYTKSGMILCRNDYIRLFGNSGACSACGQSIPASELVMRAQGNVYHLKCFTCSTCRNRLVPGDRFHYINGSLFCEHDRPTALINGHLNSLQSNPLLPDQKVCSpecifications
| LIM domain only 4 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_006769 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3B4 | |
| Mouse monoclonal antibody raised against a full length recombinant LMO4. | |
| LMO4 | |
| LMO4 | |
| LMO4 (NP_006760, 57 a.a. ∼ 165 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 8543 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction