missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM domain only 3 (rhombotin-like 2), Mouse, Clone: 4B9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LMO3.
Brand: Abnova H00055885-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: MRAKDNVYHLDCFACQLCNQRFCVGDKFFLKNNMILCQTDYEEGLMKEGYAPQVRSpecifications
| LIM domain only 3 (rhombotin-like 2) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_018640 | |
| LMO3 | |
| LMO3 (NP_061110, 91 a.a. ∼ 145 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 55885 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
| ELISA, Western Blot | |
| 4B9 | |
| Mouse monoclonal antibody raised against a partial recombinant LMO3. | |
| LMO3 | |
| DAT1/MGC26081/RBTN3/RBTNL2/RHOM3/Rhom-3 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction