Learn More
LIM homeobox 6, Mouse, Clone: 3E8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LHX6.
Brand: Abnova H00026468-M05.100ug
Description
This gene encodes a member of a large protein family that contains the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator and may be involved in the control of differentiation and development of neural and lymphoid cells. Two alternatively spliced transcript variants encoding distinct isoforms have been described for this gene. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined. [provided by RefSeq
Sequence: HKKHTPQHPVPPSGAPPSRLPSALSDDIHYTPFSSPERARMVTLHGYIESQVQCGQVHCRLPYTAPPVHLKADMDGPLSNRGEKVILFQYSpecifications
| LIM homeobox 6 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_014368 | |
| LHX6 | |
| LHX6 (NP_055183, 274 a.a. ∼ 363 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 3E8 | |
| Mouse monoclonal antibody raised against a partial recombinant LHX6. | |
| LHX6 | |
| LHX6.1/MGC119542/MGC119544/MGC119545 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 26468 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.