Learn More
Lgr5 (N-Terminal) Rat anti-Human, Alexa Fluor 647, Clone: 8F2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562903
Description
Lgr5 (leucine-rich-repeat-containing G-protein-coupled receptor 5) is a seven transmembrane-domain receptor that is a target gene for Wnt and marks stem cells in the small intestine, colon, stomach, and hair follicle. Lgr5 was initially identified as a potential stem cell marker due to restricted expression of Lgr5 in the intestinal crypt and labeling of rapidly cycling cells of the colon and intestine. Using both lineage tracing and organoid culture experiments, Lgr5 positive cells are capable of generating all types of the small intestine epithelium hence indicating that Lgr5 marks stem cells of the small intestine and colon. R-spondin growth factors, which are secreted agonists of the Wnt pathway, bind Lgr5. The binding of R-spondins to Lgr5 leads to recruitment of the Frizzled/LRP Wnt receptor complex, which binds to Wnt ligands and leads to downstream Wnt signaling. Lgr5 is up-regulated in colon and ovarian cancers and has been implicated in promotion of tumor growth and metastasis.
The 8F2 monoclonal antibody recognizes an epitope in the N-terminal region of Human Lgr5.
Host Species: Rat
Clone: 8F2
Isotype: IgG2b, κ
Species Reactivity: Hu
Immunogen: Human LGR5 DNA
Flow Cytometry
Specifications
| Lgr5 (N-Terminal) | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Human LGR5 DNA | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 8F2 | |
| GPR49, GPR67, HG38 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.