missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LTB4R2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA120740
This item is not returnable.
View return policy
Description
PA1-20740 detects Leukotriene B4 Receptor 2 in Human samples.
BLTR2 has been reported in humans in peripheral blood leukocytes, mononuclear lymphocytes, and spleen. ESTs have been isolated from a broad array of human libraries, including brain, eye, fetal lung/testis/B-cell, heart, kidney, melanocyte/uterus/fetal heart, testis, tonsil, and uterus libraries.
Specifications
| LTB4R2 | |
| Polyclonal | |
| Unconjugated | |
| LTB4R2 | |
| 5830462O07Rik; BLT2; Blt-2; BLT2R; BLTR2; Hblt2; JULF2; KPG_004; Leukotriene B4 receptor 2; leukotriene B4 receptor BLT2; LTB4 receptor JULF2; LTB4-R 2; LTB4R2; LTB4-R2; LTB4-R-2; NOP9; Seven transmembrane receptor BLTR2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 56413 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9NPC1 | |
| LTB4R2 | |
| Synthetic 16 amino acid peptide from 3rd cytoplasmic domain of human LTB4R2. | |
| 25 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction