missing translation for 'onlineSavingsMsg'
Learn More
Learn More
leucine zipper-EF-hand containing transmembrane protein 1, Mouse, Clone: 6F7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant LETM1.
Brand: Abnova H00003954-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: ESKASKRLTKRVQQMIGQIDGLISQLEMDQQAGKLAPANGMPTGENVISVAELINAMKQVKHIPESKLTSLAAALDENKDGKVNIDDLVKVIELVDKEDVHISTSQVASpecifications
| leucine zipper-EF-hand containing transmembrane protein 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_012318 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 3954 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
| ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| 6F7 | |
| Mouse monoclonal antibody raised against a partial recombinant LETM1. | |
| LETM1 | |
| LETM1 | |
| LETM1 (NP_036450, 601 a.a. ∼ 708 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction