missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LCMT1 Monoclonal Antibody (4A4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA518059

Product Code. 15334331

  • £480.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

.

Perlecan is an extracellular matrix proteoglycan. It has a large core protein of 400-450 kDa and is often produced with heparan sulfate side chains. Perlecan is found in basement membranes where it contributes to the permeability characteristics, serves as a substrate for vascular cells and binds growth factors involved in vascular remodelling.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LCMT1
Monoclonal
1 mg/mL
PBS with 0.1% sodium azide
Q6P4Z6
LCMT1
6xHis mouse PPM1.
100 μL
Primary
Mouse, Rat
Antibody
IgG1
Western Blot
4A4
Unconjugated
LCMT1
[phosphatase 2A protein]-leucine-carboxy methyltransferase; [Phosphatase 2A protein]-leucine-carboxy methyltransferase 1; AL033290; CGI-68; Lcmt; Lcmt1; LCMT-1; Leucine carboxyl methyltransferase 1; PPMT1; protein phosphatase methyltransferase 1; protein-leucine O-methyltransferase
Mouse
Protein G
RUO
30949, 361643
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.