missing translation for 'onlineSavingsMsg'
Learn More

LAG1 homolog, ceramide synthase 6, Mouse, Clone: 6B8, Abnova™

Mouse monoclonal antibody raised against a partial recombinant LASS6.

Brand:  Abnova H00253782-M02.100ug

Product Code. 16192577

  • ONLINE EXCLUSIVE PRICE
    £303.00
    £274.10 / 100µg
    Save £28.90
    9% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Sequence: PCAIALNIQANGPQIAPPNAILEKVFTAITKHPDEKRLEGLSKQLDWDVRSIQRWFRQRRNQEKPSTLTR
Specifications

Specifications

LAG1 homolog, ceramide synthase 6
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_203463
LASS6
LASS6 (NP_982288, 62 a.a. ∼ 131 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Yes
253782
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
IgG2b κ
ELISA, Western Blot
6B8
Mouse monoclonal antibody raised against a partial recombinant LASS6.
LASS6
CerS6/MGC129949/MGC129950
Mouse
Affinity Purified
RUO
Primary
Human, Rat
Antibody
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.